Thermo Scientific™ LB Agar plate with 10μg/mL Streptomycin
For cellular and molecular biology applications
Supplier: Thermo Scientific™ J63541EQF
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Used for growth of Escherichia coli, plasmid and other bacterial culturesSpecifications
2°C to 8°C | |
Plate | |
15 mm |
For growth of Escherichia coli, plasmid and other bacterial cultures | |
20 Plates | |
10 cm |
For Research Use Only