Learn More
Laxco™ LED for MZS32, MZS33, and iLED-30 Series
Laxco™ Wavelength specific LED modules
Supplier: Laxco™ LMSP520
Includes: Single, 520nm LED Module
Description
Laxco™ Wavelength specific LED modules are to be used with Laxco’s ILED-30 Series illuminators and MZS32 & MZS33 Series Microscopes. Each unit is inspected in the United States by a factory qualified technician under a ridged quality control process to insure performance.
KEY FEATURES:
- Wavelength specific LED modules
- 2-Year warranty
Warranty
2 years
Chemical Identifiers
| LED Module | |
| Stereo Microscopy; Fluorescence Stereo Microscopy; Industrial Applications; Quality Control Applications | |
| LED |
| LED | |
| 50,000 Hours | |
| 5 years optical and mechanical, 1 year electrical from date of purchase |
Specifications
| LED Module | |
| Stereo Microscopy; Fluorescence Stereo Microscopy; Industrial Applications; Quality Control Applications | |
| Single, 520nm LED Module | |
| LED | |
| 1 w |
| LED | |
| AMPS-ILED-33; MZS32 Series Stereo Microscopes; MZS33 Series Stereo Microscopes | |
| 50,000 Hours | |
| 5 years optical and mechanical, 1 year electrical from date of purchase |