missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Lauroylsarcosine, Sodium Salt, Calbiochem™,

Catalog No. 4280105GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4280105GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4280105GM Supplier MilliporeSigma™ Supplier No. 4280105GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
PubChem CID 23668817
IUPAC Name sodium 2-(N-methyldodecanamido)acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥99% (HPLC)