missing translation for 'onlineSavingsMsg'
Learn More

Lauroyl Chloride 98.0+%, TCI America™

Catalog No. D0972500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0972500ML 500 mL
D097225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0972500ML Supplier TCI America Supplier No. D0972500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-16-3
Molecular Formula C12H23ClO
Molecular Weight (g/mol) 218.77
MDL Number MFCD00000740
InChI Key NQGIJDNPUZEBRU-UHFFFAOYSA-N
Synonym lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
PubChem CID 8166
IUPAC Name dodecanoyl chloride
SMILES CCCCCCCCCCCC(Cl)=O

Specifications

Color Yellow
Boiling Point 145°C
Quantity 500 mL
UN Number 3265
Formula Weight 218.77
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Lauroyl Chloride
EINECSNumber (2)-0632
TSCA Yes