missing translation for 'onlineSavingsMsg'
Learn More

Lauroyl chloride, 98%

Catalog No. AC169030500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169030500 50 g Glass bottle
AC169032500 250 g Glass bottle
AC169030010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169030500 Supplier Thermo Scientific Chemicals Supplier No. 169030500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-16-3 | C12H23ClO | 218.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-16-3
Molecular Formula C12H23ClO
Molecular Weight (g/mol) 218.77
MDL Number MFCD00000740
InChI Key NQGIJDNPUZEBRU-UHFFFAOYSA-N
Synonym lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
PubChem CID 8166
IUPAC Name dodecanoyl chloride
SMILES CCCCCCCCCCCC(Cl)=O

Specifications

Melting Point -17.0°C
Density 0.9200g/mL
Boiling Point 134.0°C to 137.0°C (11.0 mmHg)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460
Linear Formula CH3(CH2)10COCl
Quantity 50 g
Beilstein 02, 363
Specific Gravity 0.92
Solubility Information Solubility in water: reacts. Other solubilities: organic solutions
Formula Weight 218.77
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Lauroyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 203-941-7

RUO – Research Use Only