missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lauric acid, 99%

CAS: 143-07-7 | C12H24O2 | 200.32 g/mol

Supplier:  Thermo Scientific Chemicals 167280051

Quantity 5 kg
Packaging Plastic Drum

 View more versions of this product

Catalog No. AC167280051


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

143-07-7
200.32
POULHZVOKOAJMA-UHFFFAOYSA-N
3893
dodecanoic acid
C12H24O2
MFCD00002736
lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
CHEBI:30805
CCCCCCCCCCCC(=O)O

Specifications

Lauric acid
44.0°C to 46.0°C
225.0°C (100.0 mmHg)
Authentic
Plastic Drum
CH3(CH2)10CO2H
MFCD00002736
15, 5439
Solubility in water: insoluble. Other solubilities: soluble in benzene,ether and alcohol
CCCCCCCCCCCC(=O)O
200.32
CHEBI:30805
99%
143-07-7
White
156°C
98.5% min. (GC)
C12H24O2
5 kg
02, 359
lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
POULHZVOKOAJMA-UHFFFAOYSA-N
dodecanoic acid
3893
200.32
Crystalline Powder or Flakes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 205-582-1

SDS
Documents

Documents

Promotions

Promotions