Learn More
Thermo Scientific Chemicals Lauric acid, 99%
CAS: 143-07-7 | C12H24O2 | 200.32 g/mol
Supplier: Thermo Scientific Chemicals 167280010
| Quantity | 1 kg |
|---|---|
| Packaging | Plastic bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C12H24O2 | |
| MFCD00002736 | |
| lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12 | |
| CHEBI:30805 | |
| CCCCCCCCCCCC(=O)O |
Specifications
| Lauric acid | |
| 44.0°C to 46.0°C | |
| 225.0°C (100.0 mmHg) | |
| Authentic | |
| Plastic bottle | |
| CH3(CH2)10CO2H | |
| MFCD00002736 | |
| 15, 5439 | |
| Solubility in water: insoluble. Other solubilities: soluble in benzene,ether and alcohol | |
| CCCCCCCCCCCC(=O)O | |
| 200.32 | |
| CHEBI:30805 | |
| 99% |
| 143-07-7 | |
| White | |
| 156°C | |
| 98.5% min. (GC) | |
| C12H24O2 | |
| 1 kg | |
| 02, 359 | |
| lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12 | |
| POULHZVOKOAJMA-UHFFFAOYSA-N | |
| dodecanoic acid | |
| 3893 | |
| 200.32 | |
| Crystalline Powder or Flakes |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 205-582-1