Learn More
Electron Microscopy Sciences LATEX FINGER COTS SMALL
Catalog No. 50930837
Encompass
Inexpensive, expandable finger cots. Seamless. Protective dexterity without covering the entire hand. Powderless, non-rolled. 3 1/2 mil (0.0030") and 2 3/4"(40 mm) long.
Catalog No. 50930837 Supplier Electron Microscopy Sciences Supplier No. 71130
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Inexpensive, expandable finger cots. Seamless. Protective dexterity without covering the entire hand. Powderless, non-rolled. 3 1/2 mil (0.0030") and 2 3/4"(40 mm) long.