missing translation for 'onlineSavingsMsg'
Learn More

PIG™ Latching Drum Lid for Draining

Catalog No. 18366508
Change view
Click to view available options
Color:
Black
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18366508 Yellow
18366506 Black
18366507 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18366508 Supplier PIG™ Supplier No. DRM1212YW
Only null left

Latching drum lid with drain to keep drums and hazardous waste storage containers securely closed and locked.

  • For 55 gal. Steel Drums
  • Bolt-Ring
  • Latching design opens and closes with one hand for easy in-and-out drum access
  • Nitrile gasket and locking mechanism seal lid to help reduce vapor emissions and keep drum contents secure
  • Add a drum funnel screen (sold separately) for easy draining of waste liquids into open-head drums
  • Lid stays closed while filters or containers are draining to help you meet chemical waste regulations; lid stays upright when open
  • Lockable to prevent unauthorized access
  • Durable, chemical-resistant powder coat helps ensure long service life
  • Multiple lid color options let you segregate drums

Specifications

Product Type Latching Drum Lid
Color Yellow
Sterility Non-sterile
Height (English) 6.75 in.
Height (Metric) 17 cm
Diameter (English) Inner 18 in.
Diameter (Metric) Inner 45.72 cm