missing translation for 'onlineSavingsMsg'
Learn More

PIG™ Latching Drum Lid

Catalog No. 18366517
Change view
Click to view available options
Color:
Black
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18366517 Black
18366518 Red
18366519 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18366517 Supplier PIG™ Supplier No. DRM659BK
Only null left

Latching drum lid to keep drums and hazardous waste storage containers securely closed and locked.

  • For 55 gal. Steel Drums
  • Bolt-Ring
  • Latching design opens and closes with one hand for easy in-and-out drum access - keep drums covered and avoid chemical waste fines
  • Nitrile gasket and locking mechanism seal lid to help reduce vapor emissions and keep drum contents dry and secure
  • Easy-access lid closes securely to help you meet hazardous waste regulations
  • Hinge stops keep lid upright when open
  • Durable, chemical-resistant powder coat helps ensure long service life
  • Lockable latch helps prevent unauthorized access
  • Hinge pin is made for easy lid removal without loosening band around drum
  • Multiple lid color options allow you to segregate drums for safe, compliant filling

Specifications

Product Type Latching Drum Lid
Color Black
Sterility Non-sterile
Length (English) 26.75 in.
Length (Metric) 68 cm
Width (English) 23.25 in.
Width (Metric) 59 cm
Height (English) 4.375 in.
Height (Metric) 11 cm