GA International Laser & Inkjet Matte Paper Labels 4"" x 1"" / 101.6mm x 25.4mm Rectangle Orange From -29C to +120C (-20F to +248F)
Catalog No. 5000571225
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.
Catalog No.
5000571225
Supplier
GA InternationalSupplier No.
LIP40OR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.