Learn More
GA International Laser & Inkjet Matte Paper Labels 2.75"" x 2.75"" / 69.9mm x 69.9mm Rectangle White From -29C to +120C (-20F to +248F)
Catalog No. 5000571834
Encompass
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.
Catalog No. 5000571834 Supplier GA International Supplier No. LIP19WH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.