Learn More
United Scientific Supplies Laser Diffraction Kit and Accessories
Allows students create, view, and measure diffraction and interference patterns
$371.03 - $371.03
Specifications
For Use With (Equipment) Laser Diffraction Kit
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S27823
View Documents
United Scientific Supplies
LDFKIT
Kit, Laser Diffraction Complete
Each for $371.03
Only null left
Description
Description
  • Explore the mathematics behind diffraction and interference patterns
  • Sturdy base securely holds included red and green laser pointers
  • Project various patterns against bare walls
  • Includes Diffraction Grating (1), Single-Slit Slide (1), Double-Slit Slides (2), Activity Guide
Specifications
Specifications
Laser Diffraction Kit
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'