missing translation for 'onlineSavingsMsg'
Learn More

Lasalocid A Sodium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022765
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022765 1 mg
111022766 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022765 Supplier Merck Emd Millipore Supplier No. 739961MG

Lasalocid is an antibacterial agent, belonging to the carboxylic polyether group of antibiotics, produced by the fermentation of Streptomyces lasaliensis. It is commercially available as a sodium salt and is reported to be used as a coccidiostat in poultry.

Specifications

CAS 25999-20-6
Melting Point 175°C to 185°C; 180°C (decomposition) (literature)
Flash Point Not applicable
Molecular Formula C34H53NaO8
Linear Formula C34H53NaO8
Quantity 1 mg
MDL Number MFCD00078065
Synonym Lasalocid Sodium Salt
Optical Rotation [α]/D -28°C ± 3°C = 1 in. methanol
Formula Weight 612.77
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less