missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Large Steel Drum Funnel With Self-Closing Cover

Catalog No. 19822453
Change view
Click to view available options
Includes:
32 in. Flame Arrester
6 in. Flame Arrester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19822453 6 in. Flame Arrester
19822454 32 in. Flame Arrester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19822453 Supplier Justrite™ Supplier No. 08227

Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Large Steel Drum Funnel With Self-Closing Cover helps to prevent spills when transferring flammable solvents, thinners, oils, or paint wastes. This self-closing model meets NFPA1 and the International Fire Code. It shuts and locks automatically when a fusible link melts at 165°F (74°C) under fire conditions.

Safety Drum Funnel, large steel funnel with self-closing cover and 6 in. flame arrester. For use with flammables.

Specifications

Product Type Large Steel Drum Funnel
Color Yellow
For Use With (Application) Flammables
Material Galvanized Steel
Includes 6 in. Flame Arrester