Learn More
Boekel Scientific™ Large Histology Specimen Containers
Description
- Capacity: 120 mL
- Made of Non-cytotoxic Polypropylene
- Cap made of HDPE
- Graduated on three sides
- Cap and bottom of container contain gripping for opening while wearing gloves
- Nonsterile
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |