Learn More
Chemglass Inc Clamp, Three Prong Extension, Large, Dual Adjust, 1/2"" OD x 12"" Arm
Catalog No. CG9205E11
Encompass
Clamp, Three Prong Extension, Large, Dual Adjust, 1/2" OD x 12" Arm. Dual adjust nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Extension arm for both sizes is 1/2" (13mm) OD by 12" Long. The medium clamp has a 1 1/2" (38mm) maximum grip size. The large clamp has a 2 1/2" (64mm) maximum grip size. Complete with removable vinyl and fiberglass covers for easy cleaning.
Catalog No. CG9205E11 Supplier Chemglass Inc Supplier No. CG9205E11
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clamp, Three Prong Extension, Large, Dual Adjust, 1/2" OD x 12" Arm. Dual adjust nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Extension arm for both sizes is 1/2" (13mm) OD by 12" Long. The medium clamp has a 1 1/2" (38mm) maximum grip size. The large clamp has a 2 1/2" (64mm) maximum grip size. Complete with removable vinyl and fiberglass covers for easy cleaning.