missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Large Bore Female Luer with 5/16 in. ID - Polypropylene - QC

Catalog No. 01000184
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01000184 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000184

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Large bore luers offer a higher flow rates than standard luers.

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • RoHS Compliant
  • Gamma Stable
  • Lot Traceable

Warranty

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material Non-Animal Derived Polypropylene
For Use With (Application) Medical, Pharmaceutical, Industrial applications
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Large Bore Luer
Quantity 100 Pk.