missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Laplace’s Law Demonstrator
Description
- This demonstrator consists of a base with two separate parallel conductors, a large U-shaped magnet that fits between the parallel conductors, and a conducting rod attached to a plastic disc. When a current is passed through the rods, a force is exerted on the rod with the disc which causes it to roll along the parallel rods.
- Includes activity guide
- A great device for teaching Fleming's left-hand rule
Specifications
Specifications
| Material | Plastic |
| For Use With (Application) | Magnetic Demonstrations |
| Grade | 5+ |
| Product Type | LaPlace Law Demonstrator |
Warning: Cancer and reproductive harm