missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum trifluoromethanesulfonate, anhydrous, 99%
Description
Lanthanum trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes. It is an effective catalyst for the Baylis-Hillman reaction. It also catalyzes Michael additions to methyl vinyl ketone in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lanthanum trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes. It is an effective catalyst for the Baylis-Hillman reaction. It also catalyzes Michael additions to methyl vinyl ketone in water.
Solubility
Soluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with bases. It is hygroscopic in nature. Store under dry inert gas.
Lanthanum trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes. It is an effective catalyst for the Baylis-Hillman reaction. It also catalyzes Michael additions to methyl vinyl ketone in water.
Solubility
Soluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with bases. It is hygroscopic in nature. Store under dry inert gas.
Specifications
Specifications
| Linear Formula | (CF3SO3)3La |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 9009103 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 586.1 |
| Percent Purity | 99% |
| Chemical Name or Material | Lanthanum trifluoromethanesulfonate, anhydrous |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only