missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)

Form: Approx.0.20g/mm, Cast rod, diameter is approximate | CAS: 7439-91-0 | La | 138.91 g/mol

Supplier:  Thermo Scientific Chemicals 040292DM

 View more versions of this product

Catalog No. AA40292DM

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver-White
Rare Earth Oxides Content
Silvery-white rod
99.9% (REO)
La
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
[La]
138.91
CHEBI:33336
Lanthanum
7439-91-0
100 mm
Cast Rod
MFCD00011066
FZLIPJUXYLNCLC-UHFFFAOYSA-N
lanthanum
23926

Chemical Identifiers

7439-91-0
138.91
FZLIPJUXYLNCLC-UHFFFAOYSA-N
23926
lanthanum
La
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
CHEBI:33336
[La]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 231-099-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only