missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)

CAS: 7439-91-0 | La | 138.91 g/mol

Supplier:  Thermo Scientific Chemicals 040293DM

 View more versions of this product

Catalog No. AA40293DM

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Silver to Gray
12.7mm (0.5 in.) dia.
100 mm
≈0.80g/mm
(metals basis excluding Ta)
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
[La]
138.91
CHEBI:33336
Lanthanum rod
7439-91-0
Mylar under Argon
Cast Rod
La
UN3208
FZLIPJUXYLNCLC-UHFFFAOYSA-N
lanthanum
23926
99.9%

Chemical Identifiers

7439-91-0
138.91
FZLIPJUXYLNCLC-UHFFFAOYSA-N
23926
lanthanum
La
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
CHEBI:33336
[La]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

EINECSNumber : 231-099-0

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only