missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum pieces, 99.9% (REO)

Catalog No. p-4781952
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0017514 25 g
AA0017522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0017514 Supplier Thermo Scientific Chemicals Supplier No. 00017514
Only null left

CAS: 7439-91-0 | La | 138.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-91-0
Molecular Formula La
Molecular Weight (g/mol) 138.91
MDL Number MFCD00011066
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
PubChem CID 23926
ChEBI CHEBI:33336
IUPAC Name lanthanum
SMILES [La]

Specifications

Color Silver-White
Appearance Silvery-white pieces
Density 6.17g/cm3 at 20°C
Quantity 25 g
Packaging Under Mineral Oil
Assay Percent Range 99.9% (REO)
Chemical Name or Material Lanthanum, Rare Earth Oxides Content
Hazard Category H228
Hazard Statement Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-099-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only