missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lanthanum Oxide, Reagent Grade, LabChem™
Supplier: LabChem LC160409
Specifications
4,200°C | |
White | |
100 | |
100 g | |
Soluble in water | |
Reagent | |
Poly Bottle | |
Fume; Carbon monoxide; Carbon dioxide |
Lanthanum Oxide | |
1312-81-8 | |
Powder | |
La2O3 | |
325.81 | |
Passes Test | |
6.51g/cm3 | |
6.51g/cm3 |
Chemical Identifiers
1312-81-8 |
Safety and Handling
GHS H Statement
Causes eye irritation.
GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature