missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Oxide, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5390763
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003315 100 g Wide Mouth Amber Glass Bottle
02003316 250 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003315 Supplier Avantor Supplier No. P35105
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3

Specifications

Quantity 100 g
Formula Weight 325.81
Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 6.51kg
Chemical Name or Material Lanthanum Oxide