Learn More
Lanthanum(III) phosphate hydrate, 99.99% (REO)
CAS: 14913-14-5 | LaO4P | 233.88 g/mol
Supplier: Thermo Scientific Chemicals 04199014
Description
It is an active pharmacetical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lanthanum(III) phosphate hydrate | |
| 14913-14-5 | |
| 25 g | |
| LaO4P | |
| Insoluble in water. | |
| [La+3].[O-]P([O-])([O-])=O | |
| 233.88 | |
| 99.99% |
| White | |
| Lump | |
| (REO) | |
| MFCD00150248 | |
| LQFNMFDUAPEJRY-UHFFFAOYSA-K | |
| lanthanum(3+) phosphate | |
| 233.88 (Anhydrous) | |
| Odorless |
Chemical Identifiers
| 14913-14-5 | |
| 233.88 | |
| LQFNMFDUAPEJRY-UHFFFAOYSA-K | |
| [La+3].[O-]P([O-])([O-])=O |
| LaO4P | |
| MFCD00150248 | |
| lanthanum(3+) phosphate |
Safety and Handling
EINECSNumber : 237-419-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only