missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) phosphate hydrate, 99.99% (REO)

Catalog No. AA4199006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4199006 5 g
AA4199014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4199006 Supplier Thermo Scientific Chemicals Supplier No. 04199006
Only null left

CAS: 14913-14-5 | LaO4P | 233.88 g/mol

It is an active pharmacetical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14913-14-5
Molecular Formula LaO4P
Molecular Weight (g/mol) 233.88
MDL Number MFCD00150248
InChI Key LQFNMFDUAPEJRY-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) phosphate
SMILES [La+3].[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Lump
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 233.88 (Anhydrous)
Percent Purity 99.99%
Odor Odorless
Assay (REO)
Chemical Name or Material Lanthanum(III) phosphate hydrate
EINECSNumber 237-419-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only