Learn More
Lanthanum(III) perchlorate hexahydrate
CAS: 14017-46-0 | Cl3H12LaO18 | 545.33 g/mol
$92.68 - $240.08
Chemical Identifiers
| CAS | 14017-46-0 |
|---|---|
| Molecular Formula | Cl3H12LaO18 |
| Molecular Weight (g/mol) | 545.33 |
| MDL Number | MFCD00150245 |
| InChI Key | DCEUOMQVLPJQAI-UHFFFAOYSA-K |
| Synonym | lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution |
| PubChem CID | 166977 |
| IUPAC Name | lanthanum(3+);triperchlorate |
| SMILES | O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Description
Lanthanum(III) perchlorate hexahydrate participates in the preparation of the Schiff base complexes of lanthanide (III) ions. When hydrated lanthanum perchlorate in ethanolic solution is treated with an ethanolic solution of terpyridine, [La(terpy)3](CIO4)3 is deposited as very small crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14017-46-0 | |
| 545.33 | |
| DCEUOMQVLPJQAI-UHFFFAOYSA-K | |
| 166977 | |
| O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
| Cl3H12LaO18 | |
| MFCD00150245 | |
| lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution | |
| lanthanum(3+);triperchlorate |
Specifications
| 14017-46-0 | |
| 25 g | |
| MFCD00150245 | |
| lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution | |
| DCEUOMQVLPJQAI-UHFFFAOYSA-K | |
| lanthanum(3+);triperchlorate | |
| 166977 | |
| Odorless | |
| Lanthanum(III) perchlorate hexahydrate |
| Crystalline | |
| Cl3H12LaO18 | |
| UN1481 | |
| Easily soluble in water. | |
| O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
| 545.33 | |
| 545.35 (437.26 Anhydrous) | |
| Hygroscopic |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 237-835-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only