missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) Oxide 99.9+%, TCI America™

Catalog No. L0235500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0235500G 500 g
L023525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0235500G Supplier TCI America Supplier No. L0235500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1312-81-8 | La2O3 | 325.81 g/mol

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
MDL Number MFCD00011071

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range >99.9%
Merck Index 5363
Solubility Information Insoluble in Water
Molecular Weight (g/mol) 325.81 g/moL
Formula Weight 325.81
Percent Purity ≥99.9%
Manufacturing Origin USA
Appearance White to Almost White Powder to Crystal
Chemical Name or Material Lanthanum(III) Oxide
Show More Show Less
EINECSNumber (1)-0757
RTECSNumber OE5330000
TSCA Yes