missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) Oxide 99.9+%, TCI America™

Catalog No. L023525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L023525G 25 g
L0235500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L023525G Supplier TCI America Supplier No. L023525G
Only null left

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
MDL Number MFCD00011071

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 325.81
Percent Purity ≥99.9%
Chemical Name or Material Lanthanum(III) Oxide
EINECSNumber (1)-0757
RTECSNumber OE5330000
TSCA Yes