missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lanthanum(III) oxide, REacton™, 99.9% (REO)

Catalog No. AA11272A4
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11272A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA11272A4 Supplier Thermo Scientific™ Supplier No. 11272A4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Boiling Point 4200°
Melting Point 2307°
CAS 1312-81-8
Physical Form Solid
Quantity 2.5 kg
Molecular Formula La{2}O{3}
MDL Number MFCD00011071
Merck Index 14,5363
Synonym Lanthanum trioxide;Lanthanum sesquioxide
Formula Weight 325.82
Percent Purity 99.9%
Density 6.51
Show More Show Less