missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) oxide, For AAS, ≥99.9%, MilliporeSigma™ Supelco™
Lanthanum (III) oxide is an alkaline earth oxide, and is a diamagnetic, isostructural analog of neodymium oxide. It is also an efficient recyclable catalyst, and can be used in coupling various aryl halides with aromatic/alkyl thiols.
Supplier: Merck Emd Millipore 289205250G
Specifications
| 1312-81-8 | |
| 250 g | |
| MFCD00011071 | |
| ≥99.9% (trace metals basis); ≥99.9% | |
| Poly Bottle |
| Powder | |
| La2O3 | |
| 325.81 | |
| AAS | |
| 6.51 g/mL (at 25°C) |
Chemical Identifiers
| MFCD00011071 |
Safety and Handling
RTECSNumber : OE5330000