missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) Oxide, MilliporeSigma™

Catalog No. M1122200100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1122200100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1122200100 Supplier Technidata Supplier No. 1.12220.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity Lanthanum(III) oxide for general laboratory use.

  • Lanthanum Oxide with ignited basis substance packed in 100 gm plastic bottle and stored under room temperature

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
Synonym Lanthanum III Oxide

Specifications

Physical Form Solid
Quantity 100 g
Formula Weight 325.82
Odor Odorless
Packaging Plastic bottle
Product Line EMPLURA™
Appearance White solid
Vapor Pressure Low
Chemical Name or Material Lanthanum(III) oxide