missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, 99.9% (metals basis)

For vacuum deposition | CAS: 1312-81-8

$337.18 - $337.18

Chemical Identifiers

CAS 1312-81-8
MDL Number MFCD00011071
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4164418
View Documents
Thermo Scientific Chemicals
04164418
50 g
Each for $337.18
Only null left
 
Description

Description

Lanthanum(III) oxide is used for vacuum deposition. It is also acts as a precursor to LAMOX fast ion conductors. It is also employed in the manufacture of special alloys, optical glass and refractory materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1312-81-8
MFCD00011071

Specifications

4200°C
1312-81-8
50 g
14,5363
325.82
Odorless
6.51 g/mL
(metals basis)
2307°C
Sintered Lump
MFCD00011071
Insoluble in water; Soluble in dilute mineral acids
99.9%
Hygroscopic
Lanthanum(III) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 215-200-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only