missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, 91%

Catalog No. AC413021000
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC413021000 100 g
AC413022500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413021000 Supplier Thermo Scientific Chemicals Supplier No. 413021000
Only null left

Lanthanum(III) oxide, 91%, La2O3, CAS Number-1312-81-8, 100g, 89% min, 4200.0 deg.C, White to Gray, 215-200-5, 325.82, HYGROSCOPIC, GHS H Statement: Causes serious eye irritation. | CAS: 1312-81-8 | La2O3 | 325.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011071

Specifications

Boiling Point 4200.0°C
Melting Point 2315.0°C
Color White to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 89% min
Linear Formula La2O3
Merck Index 15, 5414
Solubility Information Solubility in water: insoluble.
Formula Weight 325.82
Percent Purity 91%
Packaging Glass bottle
Chemical Name or Material Lanthanum(III) oxide, 91%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-200-5
RTECSNumber OE5330000
TSCA TSCA

RUO – Research Use Only