missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) nitrate hexahydrate, Honeywell Fluka™

Catalog No. p-7072242
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
6000884 100 g
6000885 500 g
2 options

Catalog No. 6000884

Supplier: Honeywell Chemicals 61520100G

Only null left

Puriss. p.a.,≥99.0% (titration)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
PubChem CID 16211478
IUPAC Name lanthanum(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Quantity 100 g
Linear Formula La(NO3)3 · 6H2O
UN Number UN1477
Formula Weight 433.01g/mol
Percent Purity ≥99% (titration)
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Hazard Statement P220-P261-P305 + P351 + P338
RTECSNumber OE5250000