missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) fluoride, anhydrous, 99.9% (REO)

Catalog No. p-7051517
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
Catalog No. Quantity
AA4141418 50 g
1 options

Catalog No. AA4141418

Supplier: Thermo Scientific Chemicals 04141418

Only null left

For vacuum deposition | CAS: 13709-38-1 | F3La | 195.90 g/mol

Lanthanum fluoride, anhydrous is employed extensively in the preparation and study of fluoride glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13709-38-1
Molecular Formula F3La
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011073
InChI Key BYMUNNMMXKDFEZ-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trifluoride
SMILES [F-].[F-].[F-].[La+3]

Specifications

Melting Point 1493°C
Physical Form Pieces
Quantity 50 g
Linear Formula LaF3
Solubility Information Soluble in strong mineral acids. Insoluble in water.
Formula Weight 195.9
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 5.936 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-252-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only