Learn More
Lanthanum(III) carbonate hydrate, 99.9% (REO)
CAS: 54451-24-0 | C3La2O9 | 457.84 g/mol
$45.90 - $444.86
Chemical Identifiers
| CAS | 54451-24-0 |
|---|---|
| Molecular Formula | C3La2O9 |
| Molecular Weight (g/mol) | 457.84 |
| MDL Number | MFCD00011069 |
| InChI Key | NZPIUJUFIFZSPW-UHFFFAOYSA-H |
| IUPAC Name | dilanthanum(3+) tricarbonate |
| SMILES | [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1663914
|
Thermo Scientific Chemicals
01663914 |
25 g |
Each for $45.90
|
|
|||||
|
AA1663922
|
Thermo Scientific Chemicals
01663922 |
100 g |
Each for $108.92
|
|
|||||
|
AA1663936
|
Thermo Scientific Chemicals
01663936 |
500 g |
Each for $444.86
|
|
|||||
Description
It is used in pharmaceuticals & used in medicine as a phosphate binder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 54451-24-0 | |
| 457.84 | |
| NZPIUJUFIFZSPW-UHFFFAOYSA-H | |
| [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
| C3La2O9 | |
| MFCD00011069 | |
| dilanthanum(3+) tricarbonate |
Specifications
| 54451-24-0 | |
| 25 g | |
| MFCD00011069 | |
| Insoluble in water. | |
| [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O | |
| 457.84 | |
| 99.9% | |
| 2.6 g/mL to 2.7 g/mL | |
| (REO) |
| Powder | |
| C3La2O9 | |
| 14,5364 | |
| NZPIUJUFIFZSPW-UHFFFAOYSA-H | |
| dilanthanum(3+) tricarbonate | |
| 457.85 (Anhydrous) | |
| Hygroscopic | |
| Lanthanum(III) carbonate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-599-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only