Learn More
Lanthanum foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
$605.49 - $605.49
Chemical Identifiers
| CAS | 7439-91-0 |
|---|---|
| Molecular Formula | La |
| Molecular Weight (g/mol) | 138.91 |
| MDL Number | MFCD00011066 |
| InChI Key | FZLIPJUXYLNCLC-UHFFFAOYSA-N |
| Synonym | in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation |
| PubChem CID | 23926 |
| ChEBI | CHEBI:33336 |
| IUPAC Name | lanthanum |
| SMILES | [La] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
| 7439-91-0 | |
| 138.91 | |
| FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
| 23926 | |
| lanthanum |
| La | |
| MFCD00011066 | |
| in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
| CHEBI:33336 | |
| [La] |
Specifications
| Gray to Silver | |
| 25 x 25 mm | |
| ≈0.39g/25x25mm | |
| La | |
| UN3208 | |
| FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
| lanthanum | |
| 23926 | |
| 99.9% | |
| (REO) |
| 7439-91-0 | |
| Mylar under Argon | |
| Foil | |
| MFCD00011066 | |
| in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
| [La] | |
| 138.91 | |
| CHEBI:33336 | |
| Lanthanum foil, 0.1mm (0.004 in.) thick |
Safety and Handling
GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
EINECSNumber : 231-099-0
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only