missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Thermo Scientific Chemicals

100 Orientation

$1,073.30 - $1,073.30

Chemical Identifiers

CAS 12003-65-5
Molecular Formula LaAlO3
MDL Number MFCD00144550
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38501KU
View Documents
Thermo Scientific Chemicals
038501KU
5 Ea. N/A
Only null left
 
Description

Description

Used in epitaxial thin films, non-epitaxial thin films and substrate. Mainly used in making optical glass parts and optical fibers for electronic industry. Also often used in ceramic industry and catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12003-65-5
LaAlO3
MFCD00144550

Specifications

Brown-Yellow
Plate
LaAlO3
Soluble in phosphorous acid (at >150°C). Insoluble in mineral acid ( at 25°C) and water.
Lanthanum Aluminum Oxide Substrate, Polished One Side
12003-65-5
5 Ea.
MFCD00144550
Odorless
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only