missing translation for 'onlineSavingsMsg'
Learn More

Lanolin, USP, Spectrum™ Chemical

Catalog No. p-7157689
Change view
Click to view available options
Quantity:
430 g
6 x 430 g
18 kg
180 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603707 430 g Poly Bottle
18603708 6 x 430 g Poly Bottle
18603709 18 kg Poly Pail
18603710 180 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603707 Supplier Spectrum Chemical Mfg Cor Supplier No. LA109430GM
Only null left

LA109, 8006-54-0

Spectrum™ Chemical Lanolin, USP is used as a moisturizer to treat or prevent dry, rough, scaly, itchy skin and minor skin irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8006-54-0

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Melting Range 38°-44°C
Packaging Poly Bottle
Quantity 430 g
Grade USP