Learn More
LaMotte™ Turbidity Test Kit
Description
- Readings are made by using a standard turbidity reagent to match the turbidity of a water sample
- Measurements are expressed in Jackson Turbidity Units (JTU)
- Reads from 0-200 JTU in 5 JTU steps
Specifications
Specifications
| Volume (Metric) | 60 mL |
| Range | Reads from 0-200 JTU in 5 JTU steps |
| Type | Turbidity Refill Reagent |