Learn More
LaMotte™ Calcium Hardness Test Kit
Because calcium and magnesium are both major components of the total hardness of water, measuring hardness by itself may not reveal enough.
Supplier:  LaMotte™ 360901
Catalog No. S94973
Only null left
Description
Description
  • Recirculating aquaculture systems need calcium levels greater than 50 ppm to maintain a fully functional biofilter
  • Testing takes 2 minutes
  • 0 to 200 ppm
Specifications
Specifications
To measure total hardness of water
50
0 to 200ppm
Calcium Hardness Test Kit
Product Suggestions
Product Suggestions