missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ SMART 3™ Colorimeter Water Quality Analyzer

Catalog No. 03920552
Change view
Click to view available options
Wavelength Range:
428, 520, 568, 635 nm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength Range
03920552 428, 520, 568, 635 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03920552 Supplier LaMotte™ Supplier No. LAM1910

Includes

Lithium ion rechargeable battery and USB wall charger, USB cable, four sample tubes, instrument manual including test procedures and quick start guide

Multitest colorimeter measures numerous water quality parameters with laboratory accuracy and the convenience of a portable, handheld system

Simple menu-driven operation displays results in concentration (ppm) or absorbance.

  • Waterproof to IP67
  • More than 75 preprogrammed tests in library and 25 user-entered tests
  • Choose any of four wavelengths (see Specifications) to determine absorbance of a sample at a desired wavelength—proper wavelength selection is assured, since all optical filters are included and automatically selected
  • Autoblank and auto-off
  • Using optional SMARTLink 3 Program and cable (order online), the SMART 3 Colorimeter can interface with any DOS-based computer program to send test data to a database or ASCII delimited file

Order Info

Net weight: 11 oz. (311.8g). Dimensions: 6L x 3.25W x 2.5 in. H (15 x 8 x 5.5cm).

Compliance

CE marked

Specifications

Accuracy ±0.005 AU at 1 AU
Certifications/Compliance CE, EPA
Connections Mini B USB
Display Backlit LCD
Dimensions (L x W x H) 3.5 x 7.5 x 2.5 in. (8.84 x 19.05 x 6.35 cm)
IP Rating IP67
Languages Seven user-selectable languages
Light Source LED
Voltage 3.7 V
Wavelength Range 428, 520, 568, 635 nm
For Use With (Application) Industrial water boiling and cooling, water and wastewater
Height (English) 2.5 in.
Height (Metric) 6.35 cm
Includes Lithium ion rechargeable battery and USB wall charger, USB cable, four sample tubes, instrument manual including test procedures and quick start guide
Length (English) 3.5 in.
Length (Metric) 8.84 cm
Pathlength 10 mm
Resolution Determined by reagent system
Warranty 2 Years
Weight (English) 13 oz.
Weight (Metric) 362 g
Width (English) 7.5 in.
Width (Metric) 19.05 cm
Show More Show Less