missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Reagent Refills

Catalog No. p-4398642
Change view
Click to view available options
Type:
Mixed Acid
Volume (English):
2.02 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Volume (English)
22278671 Mixed Acid 2.02 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22278671 Supplier LaMotte™ Supplier No. V6278CNH
Only null left

Testing supplies for the discerning scientist

Refills for sulfide tests, potassium iodide, chlorine, ferroine indicators and more

Specifications

Volume (Metric) 60 mL
Storage Requirements 25°C
Type Mixed Acid
Volume (English) 2.02 oz.