missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ Peracetic Acid Test Kit
Description
- Range/Sensitivity: 1 drop = 50ppm Peroxide, 1 drop = 15ppm Peracetic Acid
- No. of tests: 50
- No. of reagents: 5
Specifications
Specifications
| For Use With (Application) | Industrial water boiling and cooling, water and wastewater |
| Includes | 30mL sulfuric acid, 15mL ferroin indicator, 2 x 30mL hydrogen peroxide titrant, 30mL potassium iodide 20% solution, 60mL peracetic acid titrant, test tube, 5 to 10 to 25mL, plastic, with cap, 0.5mL plastic pipet |
| Quantity | 50 Tests |
| Test Method | Dropper bottle |
| Type | Peracetic Acid Test Kit |