Learn More
LaMotte™ Total Coliform Test Kit - Recreational Water
Description
- The water sample is placed in a tube containing a reagent tablet and incubated at room temperature for 48 hours
- The tube is then examined for positive indicators of coliform bacteria such as color change and formation of gas bubbles
- Each kit contains 44 capped tubes each containing a Coliform Test Tablet, and a set of laminated color chart and instruction flashcards
Specifications
Specifications
| Volume (Metric) | 100 mL |
| For Use With (Application) | Water monitoring educations |
| Includes | 44 capped tubes each containing a Coliform Test Tablet, and a set of laminated color chart and instruction flashcards. |
| No. of Tests | 44 |
| Type | Total Coliform Test Kit |