Learn More
LaMotte™ Dissolved Oxygen Test Kit
Catalog No. 22307857
Change view
Click to view available options
Range:
0 to 10ppm
Catalog No. Range
22307857 0 to 10ppm
1 options
Catalog No. 22307857 Supplier LaMotte™ Supplier No. 586001
Only null left
Direct Reading Titrator - Test uses a 1.0mL microburet calibrated to allow direct reading of the test result within a specific range
  • Dissolved Oxygen - Test uses the azide modification of the Winkler method: oxygen in presence of a strong alkali oxidizes manganese which reacts with iodide to form iodine, iodine reacts with thiosulfate in presence of starch (to enhance endpoint)
  • Range: 0 to 10ppm
  • 50 tests at 10ppm per kit
Specifications
For Use With (Application) Measures for dissolved oxygen levels in food and beverage, environmental education, industrial water boiling and cooling, laundry, sanitation, water and wastewater
Includes 30mL manganous sulfate solution, 30mL alkaline potassium iodide azide, 30mL sulfuric acid, 60mL 0.025N sodium thiosulfate, 30mL starch indicator solution, direct reading titrator, test tube with cap, 60mL glass water sampling bottle
No. of Tests 50
Range 0 to 10ppm
Test Method Azide modification of Winkler titration
Type Dissolved Oxygen Test Kit