Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems
SMART 3™ Colorimeter Reagent Systems for use in water quality testing and environmental studies.
Supplier: LaMotte™ 402501
Description
- Reagents for use in water testing and environmental studies.
Chemical Identifiers
| SMART 3 Colorimeter |
Specifications
| SMART 3 Colorimeter | |
| 0.0 to 70 ppm | |
| Phosphorus Total HR Reagent |
| 25 | |
| Molybdovanadate or Digestion |