missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine 98.0+%, TCI America™

Catalog No. p-7131889
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02411G 1 g
L02415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02411G Supplier TCI America Supplier No. L02411G
Only null left

Chemical Identifiers

CAS 84057-84-1
Molecular Formula C9H7Cl2N5
Molecular Weight (g/mol) 256.09
MDL Number MFCD00865333
InChI Key PYZRQGJRPPTADH-UHFFFAOYSA-N
Synonym lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
PubChem CID 3878
ChEBI CHEBI:6367
IUPAC Name 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
SMILES NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 256.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lamotrigine
RTECSNumber XY5850700
TSCA No
Recommended Storage Refrigerator